Läs mer
3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate, Mikromol™
Produktkod. 18042000
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
18042000 100 mg
1 options
Produktkod. 18042000 Leverantör Mikromol Leverantörsnummer MM0383.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dehydro Amlodipine
CAS 113994-41-5
Analyt- eller komponentnamn Dehydro Amlodipine
Frakt skick Ice pack (-20°C)
Molekylformel C20 H23 Cl N2 O5
Certifikat och efterlevnad ISO 9001
Synonym Amlodipine Besilate Imp. D (EP), Amlodipine Imp. D (EP), Amlodipine USP Related Compound A, Amlodipine USP RC A, 3-Ethyl 5-Methyl 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate, Amlodipine Related Compound A (USP) as free base, Amlodipine Related Compound A, Amlodipine Besilate Impurity D, Amlodipine Impurity D
InChI formel InChI=1S/C20H23ClN2O5/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21/h5-8H,4,9-11,22H2,1-3H3
LEDER CCOC(=O)c1c(COCCN)nc(C)c(C(=O)OC)c1c2ccccc2Cl
IUPAC-namn 3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate
Molekylvikt (g/mol) 406.86
Kvantitet 100 mg
Formel vikt 406.1295 g/mol