missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
3-Etyl 5-metyl (4RS)-4-(2-klorofenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)etoxi]metyl]-6-metyl-1,4-dihydropyridin-3,5-dikarboxylat, LoGiCal
Beskrivning
3-Etyl 5-metyl (4RS)-4-(2-klorofenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)etoxi]metyl]-6-metyl-1,4-dihydropyridin-3,5-dikarboxylat
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-([2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 88150-62-3 |
| Analyt- eller komponentnamn | 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-([2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Frakt skick | Rumstemperatur |
| Molekylformel | C28 H27 Cl N2 O7 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. A (EP), Amlodipine Imp. A (EP), Amlodipine USP Related Compound D, Amlodipine USP RC D, 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-Methyl (4 RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| InChI formel | InChI=1 S/C28H27ClN2O7/c1-4-38-28(35)24-21(15-37-14-13-31-25(32)17-9-5-6-10-18(17)26(31)33)30-16(2)22(27(34)36-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,30 H,4,13-15H2,1-3H3 |
| LEDER | CCOC(=O)C1=C(COCCN2C(=O)c3ccccc3C2=O)NC(=C(C1c4ccccc4Cl)C(=O)OC)C |
| IUPAC-namn | 3-O-etyl 5-O-metyl 4-(2-klorofenyl)-2-[2-(1,3-dioxoisoindol-2-yl)etoxymetyl]-6-metyl-1,4-dihydropyridin-3,5-dikarboxylat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?