Läs mer
3-Ethyl Haloperidol, TRC
Produktkod. 30378376
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378376 100 mg
30371284 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30378376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE918700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391052-87-1
Molekylformel C23 H27 Cl F N O2
Molekylvikt (g/mol) 403.92
Synonym 4-(4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl)-1-(3-ethyl-4-fluorophenyl)butan-1-one
IUPAC-namn 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidyl]-1-(3-ethyl-4-fluoro-phenyl)butan-1-one
LEDER CCc1cc(ccc1F)C(=O)CCCN2CCC(O)(CC2)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material Haloperidol EP Impurity C
Kvantitet 100 mg
InChI formel InChI=1S/C23H27ClFNO2/c1-2-17-16-18(5-10-21(17)25)22(27)4-3-13-26-14-11-23(28,12-15-26)19-6-8-20(24)9-7-19/h5-10,16,28H,2-4,11-15H2,1H3
Formel vikt 403.1714