Läs mer
3-Ethylhexane, TRC
Produktkod. 30388043
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30388043 50 mg
30370572 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30388043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91876550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 619-99-8
Molekylformel C8 H18
Molekylvikt (g/mol) 114.23
Synonym Hexane, 3-ethyl-, 3-Ethylhexane, NSC 74175
IUPAC-namn 3-ethylhexane
LEDER CCCC(CC)CC
Specifikationer
Kemiskt namn eller material 3-Ethylhexane
Kvantitet 50 mg
InChI formel InChI=1S/C8H18/c1-4-7-8(5-2)6-3/h8H,4-7H2,1-3H3
Formel vikt 114.1409