Learn More
3-Ethynylthiophene, 96%, Thermo Scientific Chemicals
Catalog Number p-4228655
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
67237-53-0
C6H4S
108.16
MJHLPKWONJUCFK-UHFFFAOYSA-N
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b
3548422
3-ethynylthiophene
C#CC1=CSC=C1
67237-53-0
C6H4S
108.16
MJHLPKWONJUCFK-UHFFFAOYSA-N
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b
3548422
3-ethynylthiophene
C#CC1=CSC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Orange
Density 1.0980g/mL
Boiling Point 152°C to 154°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.098
Formula Weight 108.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Ethynylthiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Ethynylthiophene, 96%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.