Läs mer
3-Ferrocenoylpropionic Acid, TRC
Produktkod. 30406407
Change view
Kvantitet:
500 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30406407 2.5 g 2.5g
30392855 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30406407 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF2858052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Ferrocenoylpropionic Acid
CAS 1291-72-1
Molekylformel C14H14FeO3
Kvantitet 2.5 g
LEDER C1=C[CH]C(=C1)C(=O)CCC(=O)O.C1=CCC=C1.[Fe]
Molekylvikt (g/mol) 286.1