missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxy-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number 15471989
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
375368-80-2
C7H8FNO
141.15
MFCD03095083
DAQMWRIPXIPMAA-UHFFFAOYSA-N
3-fluoro-2-methoxy-6-picoline, 2-methoxy-3-fluoro-6-methylpyridine, 3-fluoro-2-methoxy-6-methyl-pyridine, pubchem6585, ksc914i9l, abbypharma ap-15-5217, pyridine, 3-fluoro-2-methoxy-6-methyl, pyridine, 3-fluoro-2-methoxy-6-methyl-9ci
24820319
3-fluoro-2-methoxy-6-methylpyridine
COC1=C(F)C=CC(C)=N1
This item is not returnable. View return policy
375368-80-2
C7H8FNO
141.15
MFCD03095083
DAQMWRIPXIPMAA-UHFFFAOYSA-N
3-fluoro-2-methoxy-6-picoline, 2-methoxy-3-fluoro-6-methylpyridine, 3-fluoro-2-methoxy-6-methyl-pyridine, pubchem6585, ksc914i9l, abbypharma ap-15-5217, pyridine, 3-fluoro-2-methoxy-6-methyl, pyridine, 3-fluoro-2-methoxy-6-methyl-9ci
24820319
3-fluoro-2-methoxy-6-methylpyridine
COC1=C(F)C=CC(C)=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Quantity 1 g
Formula Weight 141.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methoxy-6-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'