missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15457716
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1192548-05-2
C13H15BF4O2
290.06
MFCD16996330
KVMWQTWKCBGEBD-UHFFFAOYSA-N
2-3-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-2-trifluoromethylphenylboronic acid pinacol ester, 3-fluoro-2-trifluoromethyl benzeneboronic acid pinacol ester, 3-fluoro-2-trifluoromethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl
57993875
2-[3-fluoro-2-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(C(F)=CC=C1)C(F)(F)F
This item is not returnable. View return policy
1192548-05-2
C13H15BF4O2
290.06
MFCD16996330
KVMWQTWKCBGEBD-UHFFFAOYSA-N
2-3-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-2-trifluoromethylphenylboronic acid pinacol ester, 3-fluoro-2-trifluoromethyl benzeneboronic acid pinacol ester, 3-fluoro-2-trifluoromethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-2-trifluoromethyl phenyl-4,4,5,5-tetramethyl
57993875
2-[3-fluoro-2-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(C(F)=CC=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 34°C to 36°C
Quantity 250 mg
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'