Learn More
3-Fluoro-4-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog Number p-4812944
Click to view available options
Quantity:
1 g
5 g
40273-47-0
C6H4FNO
125.10
MFCD06410686
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
3-fluoroisonicotinaldehyde, 3-fluoropyridine-4-carboxaldehyde, 3-fluoro-4-formylpyridine, 3-fluoro-4-pyridinecarboxaldehyde, 4-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-pyridine-4-carbaldehyde, 3-fluoro-4-pyridinecarbaldehyde, brettphospd, pubchem5137, acmc-20a1qb
2763003
3-fluoropyridine-4-carbaldehyde
FC1=C(C=O)C=CN=C1
40273-47-0
C6H4FNO
125.10
MFCD06410686
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
3-fluoroisonicotinaldehyde, 3-fluoropyridine-4-carboxaldehyde, 3-fluoro-4-formylpyridine, 3-fluoro-4-pyridinecarboxaldehyde, 4-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-pyridine-4-carbaldehyde, 3-fluoro-4-pyridinecarbaldehyde, brettphospd, pubchem5137, acmc-20a1qb
2763003
3-fluoropyridine-4-carbaldehyde
FC1=C(C=O)C=CN=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.2550g/mL
Boiling Point 70.0°C (25.0 Torr)
Flash Point 67°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.255
Formula Weight 125.1
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-pyridinecarboxaldehyde
compliance-icons
Product Identifier
  • 3-Fluoro-4-pyridinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.