missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-5-iodopyridine, 95%

Brand:  Alfa Aesar™ H64770.06

Additional Details : CAS Number : 757950-13-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15462109

  • 4203.69 SEK / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-5-iodopyridine
222.99
11107918
757950-13-3
MFCD08457780

Specifications

3-Fluoro-5-iodopyridine
Off-white
757950-13-3
95%
MFCD08457780
Light Sensitive
5g
Solid
222.99
11107918
Videos
SDS