missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62841.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15437906

  • 2156.00 SEK / 250mg


Estimated Shipment
19-10-2020
Add to basket

Specifications

Chemical Identifiers

1416367-00-4
252.092
2-3-fluoro-5-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-5-methoxyphenylboronic acid pinacol ester, 3-fluoro-5-methoxybenzeneboronic acid pinacol ester
2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C13H18BFO3
DGHRKRRFVKPHDQ-UHFFFAOYSA-N
72221149
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OC

Specifications

3-Fluoro-5-methoxybenzeneboronic acid pinacol ester
1416367-00-4
2-3-fluoro-5-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-5-methoxyphenylboronic acid pinacol ester, 3-fluoro-5-methoxybenzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OC
252.092
252.09
96%
250mg
C13H18BFO3
DGHRKRRFVKPHDQ-UHFFFAOYSA-N
2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
72221149
Solid
Videos
SDS Documents