Learn More
Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%
Brand:  Alfa Aesar™ H62937.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15488016
  • 5601.71 SEK / 1g
Estimated Shipment date
from Supplier 06-07-2018
Add to basket
Specifications
Chemical Identifiers
3-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester
290.06
MFCD18729929
5-Fluoro-3-(trifluoromethyl)phenylboronic acid pinacol ester
Specifications
3-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester
Low Melting Solid
290.06
5-Fluoro-3-(trifluoromethyl)phenylboronic acid pinacol ester
1g
96%
MFCD18729929