missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3757952
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1711-07-5
C7H4ClFO
158.56
MFCD00000670
SYVNVEGIRVXRQH-UHFFFAOYSA-N
m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
74376
3-fluorobenzoyl chloride
C1=CC(=CC(=C1)F)C(=O)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1711-07-5
C7H4ClFO
158.56
MFCD00000670
SYVNVEGIRVXRQH-UHFFFAOYSA-N
m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
74376
3-fluorobenzoyl chloride
C1=CC(=CC(=C1)F)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -30.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 189.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCl
Refractive Index 1.5260 to 1.5300
Quantity 5 mL
Beilstein 09, I, 137
Specific Gravity 1.3
Solubility Information Solubility in water: decomposes
Formula Weight 158.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'