Läs mer
3-Fluoropropan-1-ol, TRC
Produktkod. 30405511
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405511 1 g
30395628 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30405511 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5986051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Fluoropropan-1-ol
CAS 462-43-1
Molekylformel C3H7FO
Kvantitet 1 g
Synonym 3-Fluoro-1-propanol, 3-Fluoropropanol, 3-Hydroxypropyl Fluoride
InChI formel InChI=1S/C3H7FO/c4-2-1-3-5/h5H,1-3H2
LEDER OCCCF
IUPAC-namn 3-fluoropropan-1-ol
Molekylvikt (g/mol) 78.09
Formel vikt 78.0481