missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-iodopyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in the synthesis of isatogens via tolan (diphenylacetylene) intermediates.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
It is used in the synthesis of isatogens via tolan (diphenylacetylene) intermediates.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications
Melting Point | 191°C to 195°C |
Quantity | 1 g |
Beilstein | 109836 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 221 |
Percent Purity | 98% |
Chemical Name or Material | 3-Hydroxy-2-iodopyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'