Läs mer
3-Hydroxy-2-methylbutanoic Acid, TRC
Produktkod. 30012636
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012636 50 mg
30004747 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012636 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94648550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 473-86-9
Molekylformel C5 H10 O3
Molekylvikt (g/mol) 118.13
Synonym Butanoic acid, 3-hydroxy-2-methyl-, Butyric acid, 3-hydroxy-2-methyl- (6CI,8CI), 3-Hydroxy-2-methylbutanoic acid, 2-Methyl-3-hydroxybutyric acid, 3-Hydroxy-2-methylbutyric acid, Nilic acid, α-Methyl-β-hydroxybutyric acid
IUPAC-namn 3-hydroxy-2-methylbutanoic acid
LEDER CC(O)C(C)C(=O)O
Specifikationer
Kemiskt namn eller material 3-Hydroxy-2-methylbutanoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C5H10O3/c1-3(4(2)6)5(7)8/h3-4,6H,1-2H3,(H,7,8)
Formel vikt 118.063