Läs mer
3-Hydroxy-4-methoxymandelic Acid, TRC
Produktkod. 30132950
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132950 1 g
30008787 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9463451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3695-24-7
Molekylformel C9 H10 O5
Molekylvikt (g/mol) 198.17
Synonym Benzeneacetic acid, α,3-dihydroxy-4-methoxy-, Mandelic acid, 3-hydroxy-4-methoxy- (7CI,8CI), α,3-Dihydroxy-4-methoxybenzeneacetic acid, (RS)-3-Hydroxy-4-methoxymandelic acid, (±)-3-Hydroxy-4-methoxymandelic acid, 3-Hydroxy-4-methoxymandelic acid, Isovanillyl-dl-mandelic acid, Isovanilmandelic acid
IUPAC-namn 2-hydroxy-2-(3-hydroxy-4-methoxyphenyl)acetic acid
LEDER COc1ccc(cc1O)C(O)C(=O)O
Specifikationer
Kemiskt namn eller material 3-Hydroxy-4-methoxymandelic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C9H10O5/c1-14-7-3-2-5(4-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)
Formel vikt 198.0528
Procent renhet >95%