Läs mer
3-Hydroxy-4-morpholino-1,2,5-thiazole, TRC
Produktkod. 30137639
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137639 2.5 g 2.5g
17983305 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9484752.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30165-97-0
Molekylformel C6 H9 N3 O2 S
Molekylvikt (g/mol) 187.22
Synonym 4-(Morpholin-4-yl)-1,2,5-thiadiazol-3-ol, Timolol Maleate Imp. D (EP)
IUPAC-namn 4-morpholin-4-yl-1,2,5-thiadiazol-3-ol
LEDER Oc1nsnc1N2CCOCC2
Specifikationer
Kemiskt namn eller material 4-(Morpholin-4-yl)-1,2,5-thiadiazol-3-ol
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C6H9N3O2S/c10-6-5(7-12-8-6)9-1-3-11-4-2-9/h1-4H2,(H,8,10)
Formel vikt 187.0415
Procent renhet >95%