Läs mer
3-Hydroxy Carbofuran, TRC
Produktkod. 18011120
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18011120 50 mg
17999619 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18011120 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH82504050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16655-82-6
Molekylformel C12 H15 N O4
Molekylvikt (g/mol) 237.25
Synonym 3,7-Benzofurandiol, 2,3-dihydro-2,2-dimethyl-, 7-(methylcarbamate) (9CI), Carbamic acid, methyl-, 2,3-dihydro-3-hydroxy-2,2-dimethyl-7-benzofuranyl ester (8CI), 2,3-Dihydro-2,2-dimethyl-3-hydroxy-7-benzofuranyl N-methylcarbamate, 2,3-Dihydro-2,2-dimethyl-3-hydroxy-7-benzofuranyl methylcarbamate, 2,3-Dihydro-3-hydroxy-2,2-dimethyl-7-benzofuranyl methylcarbamate, 3-Hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylcarbamate, 3-Hydroxycarbofuran
IUPAC-namn (3-hydroxy-2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate
LEDER CNC(=O)Oc1cccc2C(O)C(C)(C)Oc12
Specifikationer
Kemiskt namn eller material Carbofuran-3-hydroxy
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C12H15NO4/c1-12(2)10(14)7-5-4-6-8(9(7)17-12)16-11(15)13-3/h4-6,10,14H,1-3H3,(H,13,15)
Formel vikt 237.1001
Procent renhet >95%