Läs mer
3-Hydroxy Darifenacin (Mixture of Diastereomers), TRC
Produktkod. 30405156
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30405156 1 mg
1 options
Produktkod. 30405156 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8251901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-hydroxy Darifenacin (mixture of diastereomers)
CAS 1285875-62-8
Molekylformel C28 H30 N2 O3
Kvantitet 1 mg
Synonym 3-Pyrrolidineacetamide, 1-[2-(2,3-dihydro-3-hydroxy-5-benzofuranyl)ethyl]-α,α-diphenyl-, 1-[2-(2,3-Dihydro-3-hydroxy-5-benzofuranyl)ethyl]-α,α-diphenyl-3-pyrrolidineacetamide, 2-[[1-(2,3-Dihydro-3-hydroxy-5-benzofuranyl)ethyl]pyrrolidin-3-yl]-2,2-diphenylacetamide, 3-Hydroxy Darifenacin (Mixture of Diastereomers)
InChI formel InChI=1S/C28H30N2O3/c29-27(32)28(21-7-3-1-4-8-21,22-9-5-2-6-10-22)23-14-16-30(18-23)15-13-20-11-12-26-24(17-20)25(31)19-33-26/h1-12,17,23,25,31H,13-16,18-19H2,(H2,29,32)
LEDER NC(=O)C(C1CCN(CCc2ccc3OCC(O)c3c2)C1)(c4ccccc4)c5ccccc5
IUPAC-namn 2-[1-[2-(3-hydroxy-2,3-dihydro-1-benzofuran-5-yl)ethyl]pyrrolidin-3-yl]-2,2-diphenylacetamide
Molekylvikt (g/mol) 442.55
Formel vikt 442.2256