Läs mer
3-Hydroxy Lidocaine, TRC
Produktkod. 30011281
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011281 1 mg
30009660 5 mg
30012004 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9438201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34604-55-2
Molekylformel C14 H22 N2 O2
Molekylvikt (g/mol) 250.34
Synonym Acetamide, 2-(diethylamino)-N-(3-hydroxy-2,6-dimethylphenyl)-, 2-(Diethylamino)-N-(3-hydroxy-2,6-dimethyl-phenyl)acetamide, Lidocaine-3-hydroxy
IUPAC-namn 2-(diethylamino)-N-(3-hydroxy-2,6-dimethylphenyl)acetamide
LEDER CCN(CC)CC(=O)Nc1c(C)ccc(O)c1C
Specifikationer
Kemiskt namn eller material 3-Hydroxy Lidocaine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C14H22N2O2/c1-5-16(6-2)9-13(18)15-14-10(3)7-8-12(17)11(14)4/h7-8,17H,5-6,9H2,1-4H3,(H,15,18)
Formel vikt 250.1681
Procent renhet >95%