Läs mer
3-Hydroxy Ropivacaine, TRC
Produktkod. 30137561
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137561 25 mg
30137086 5 mg
30010671 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95342025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163589-30-8
Molekylformel C17H26N2O2
Molekylvikt (g/mol) 290.4
Synonym (2S)-N-(3-Hydroxy-2,6-dimethylphenyl)-1-propyl-2-piperidinecarboxamide;
LEDER CCCN1CCCC[C@H]1C(=O)Nc2c(C)ccc(O)c2C
Specifikationer
Kemiskt namn eller material 3-Hydroxy Ropivacaine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C17H26N2O2/c1-4-10-19-11-6-5-7-14(19)17(21)18-16-12(2)8-9-15(20)13(16)3/h8-9,14,20H,4-7,10-11H2,1-3H3,(H,18,21)
Formel vikt 290.1994
Procent renhet >95%