Läs mer
3-Hydroxyanthranilic Acid, TRC
Produktkod. 30010944
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010944 1 g
30009300 100 mg
30009726 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010944 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8017901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 548-93-6
Molekylformel C7 H7 N O3
Molekylvikt (g/mol) 153.14
Synonym Benzoic acid, 2-amino-3-hydroxy-, Anthranilic acid, 3-hydroxy- (8CI), 2-Amino-3-hydroxybenzoic acid, 3-Hydroxyanthranilic acid, NSC 522891
IUPAC-namn 2-amino-3-hydroxybenzoic acid
LEDER Nc1c(O)cccc1C(=O)O
Specifikationer
Kemiskt namn eller material 3-Hydroxyanthranilic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C7H7NO3/c8-6-4(7(10)11)2-1-3-5(6)9/h1-3,9H,8H2,(H,10,11)
Formel vikt 153.0426
Procent renhet >95%