missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-144299
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
81012-99-9
C7H12Cl2N2O
211.09
MFCD00044695
ONOJPUDFIOEGCX-UHFFFAOYSA-N
3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
2724402
3-(hydrazinylmethyl)phenol;dihydrochloride
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
This item is not returnable. View return policy
81012-99-9
C7H12Cl2N2O
211.09
MFCD00044695
ONOJPUDFIOEGCX-UHFFFAOYSA-N
3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
2724402
3-(hydrazinylmethyl)phenol;dihydrochloride
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 138°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4CH2NHNH2·2HCl
Quantity 1 g
Formula Weight 211.09
Percent Purity 98%
Physical Form Adhering Powder
Chemical Name or Material 3-Hydroxybenzylhydrazine dihydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybenzylhydrazine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'