Läs mer
3-Hydroxyestra-1,3,5(10),9(11)-tetraen-17beta-yl Pentanoate (9,11-Didehydroestradiol Valerate), Mikromol™
Produktkod. 18012370
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012370 100 mg
18063360 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18012370 Leverantör Mikromol Leverantörsnummer MM0209.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Hydroxyestra-1,3,5(10),9(11)-tetraen-17beta-yl Pentanoate (9,11-Didehydroestradiol Valerate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9,11-Didehydroestradiol Valerate
CAS 95959-20-9
Analyt- eller komponentnamn 9,11-Didehydroestradiol Valerate
Frakt skick Room Temperature
Molekylformel C23 H30 O3
Certifikat och efterlevnad ISO 17025
Synonym 3-Hydroxyestra-1,3,5(10),9(11)-tetraen-17beta-yl Pentanoate, Estradiol Valerate Imp. C (EP), 9,11-Didehydroestradiol Valerate, Estradiol-9-ene valerate, Estradiol Valerate Impurity C
InChI formel InChI=1S/C23H30O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,12,14,19-21,24H,3-6,8,10-11,13H2,1-2H3/t19-,20+,21+,23+/m1/s1
LEDER CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4C3=CC[C@]12C
IUPAC-namn [(8S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molekylvikt (g/mol) 354.48
Kvantitet 100 mg
Formel vikt 354.2195 g/mol