Läs mer
3-Hydroxyglutaric Acid Dimethyl Ester, TRC
Produktkod. 30406355
Change view
Kvantitet:
2 g
Produktkod. Kvantitet
30406355 2 g
1 options
Produktkod. 30406355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9425902G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Hydroxyglutaric Acid Dimethyl Ester
CAS 7250-55-7
Molekylformel C7H12O5
Kvantitet 2 g
Synonym 3-Hydroxy-pentanedioic Acid Dimethyl Ester, Dimethyl 3-hydroxyglutarate, NSC 30047
InChI formel InChI=1S/C7H12O5/c1-11-6(9)3-5(8)4-7(10)12-2/h5,8H,3-4H2,1-2H3
LEDER COC(=O)CC(O)CC(=O)OC
IUPAC-namn dimethyl 3-hydroxypentanedioate
Molekylvikt (g/mol) 176.17
Formel vikt 176.0685