Läs mer
3-Hydroxyhexadecanoic Acid Methyl Ester, TRC
Produktkod. 17917544
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17917544 5 g
17908414 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17917544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8253805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51883-36-4
Molekylformel C17 H34 O3
Molekylvikt (g/mol) 286.45
Synonym Methyl 3-Hydroxyhexadecanoate, Methyl 3-Hydroxypalmitate, 3-OH PAME;
IUPAC-namn methyl 3-hydroxyhexadecanoate
LEDER CCCCCCCCCCCCCC(O)CC(=O)OC
Specifikationer
Kemiskt namn eller material Methyl 3-Hydroxyhexadecanoate
Kvantitet 5 g
InChI formel InChI=1S/C17H34O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-16(18)15-17(19)20-2/h16,18H,3-15H2,1-2H3
Formel vikt 286.2508