Läs mer
3-(Hydroxymethyl)-1-oxy-2,2,5,5-tetramethylpyrrolidine, TRC
Produktkod. 30402347
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30402347 25 mg
30399842 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30402347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94825525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27298-75-5
Molekylformel C9 H18 N O2
Molekylvikt (g/mol) 172.24
IUPAC-namn (1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrolidin-3-yl)methanol
LEDER CC1(C)CC(CO)C(C)(C)N1[O]
Specifikationer
Kemiskt namn eller material 3-(Hydroxymethyl)-1-oxy-2,2,5,5-tetramethylpyrrolidine
Kvantitet 25 mg
InChI formel InChI=1S/C9H18NO2/c1-8(2)5-7(6-11)9(3,4)10(8)12/h7,11H,5-6H2,1-4H3
Formel vikt 172.1338