Läs mer
3-(Hydroxymethyl)Benzamide (~85%), TRC
Produktkod. 30394088
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30394088 25 mg
30405099 250 mg
30402625 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30394088 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH12692025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126926-34-9
Molekylformel C8H9NO2
Molekylvikt (g/mol) 151.16
Synonym 3-(Hydroxymethyl)benzamide
IUPAC-namn 3-(hydroxymethyl)benzamide
LEDER NC(C1=CC=CC(CO)=C1)=O
Specifikationer
Kemiskt namn eller material 3-(Hydroxymethyl)Benzamide
Kvantitet 25 mg
InChI formel InChI=1S/C8H9NO2/c9-8(11)7-3-1-2-6(4-7)5-10/h1-4,10H,5H2,(H2,9,11)
Formel vikt 151.0633