Läs mer
3-Hydroxynaphthalene-2,7-disulfonic Acid Disodium Salt (Technical Grade), TRC
Produktkod. 30008034
Change view
Kvantitet:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008034 10 g
17979994 50 g
17918114 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008034 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94958010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 135-51-3
Molekylformel C10 H6 O7 S2 . 2 Na
Molekylvikt (g/mol) 348.26
Synonym 3-Hydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, 2-Hydroxy-3,6-naphthalenedisulfonic Acid Disodium Salt, 2-Naphthol-3,6-disulfonic Acid Disodium Salt, Disodium 2-Hydroxy-3,6-naphthalenedisulfonate, Disodium 2-Naphthol-3,6-disulfonate Disodium, 3-Hydroxy-2,7-naphthalenedisulfonate Fericon;
IUPAC-namn disodium;3-hydroxynaphthalene-2,7-disulfonate
LEDER [Na+].[Na+].Oc1cc2ccc(cc2cc1S(=O)(=O)[O-])S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Hydroxynaphthalene-2,7-disulfonic Acid Disodium Salt
Kvantitet 10 g
InChI formel InChI=1S/C10H8O7S2.2Na/c11-9-4-6-1-2-8(18(12,13)14)3-7(6)5-10(9)19(15,16)17;;/h1-5,11H,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2
Formel vikt 347.935