missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-147325
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
109-00-2
C5H5NO
95.1
MFCD00006378
GRFNBEZIAWKNCO-UHFFFAOYSA-N
3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
7971
pyridin-3-ol
C1=CC(=CN=C1)O
This item is not returnable. View return policy
109-00-2
C5H5NO
95.1
MFCD00006378
GRFNBEZIAWKNCO-UHFFFAOYSA-N
3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
7971
pyridin-3-ol
C1=CC(=CN=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 123.0°C to 130.0°C
Color Beige to Brown
Boiling Point 180.0°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 21, 46
Fieser 01,486
Solubility Information Solubility in water: 33g/L water (20°C).
Formula Weight 95.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'