Läs mer
3-Hydroxytetradecanoic Acid, TRC
Produktkod. 30408166
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30408166 500 mg
1 options
Produktkod. 30408166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH956780500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Hydroxymyristic Acid
CAS 1961-72-4
Molekylformel C14 H28 O3
Kvantitet 500 mg
Synonym β-Hydroxymyristic Acid, β-Hydroxytetradecanoic Acid, (3RS)-3-Hydroxytetradecanoic Acid, (RS)-3-Hydroxytetradecanoic Acid, (±)-3-Hydroxytetradecanoic Acid, 3-Hydroxymyristic Acid
InChI formel InChI=1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17/h13,15H,2-12H2,1H3,(H,16,17)
LEDER CCCCCCCCCCCC(O)CC(=O)O
IUPAC-namn 3-hydroxytetradecanoic acid
Molekylvikt (g/mol) 244.37
Formel vikt 244.2038