Läs mer
3’-Hydroxytyrosol 3’-Glucuronide, TRC
Produktkod. 30008803
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008803 2.5 mg
30012138 1 mg
30006521 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008803 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9770302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 425408-50-0
Molekylformel C14H18O9
Molekylvikt (g/mol) 330.29
Synonym 2-Hydroxy-5-(2-hydroxyethyl)phenyl β-D-Glucopyranosiduronic Acid
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-hydroxy-5-(2-hydroxyethyl)phenoxy)tetrahydro-2H-pyran-2-carboxylic acid
LEDER O([C@@H]1O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]1O)C2=CC(CCO)=CC=C2O
Specifikationer
Kemiskt namn eller material 3'-Hydroxytyrosol 3'-Glucuronide
Kvantitet 2.5 mg
InChI formel InChI=1S/C14H18O9/c15-4-3-6-1-2-7(16)8(5-6)22-14-11(19)9(17)10(18)12(23-14)13(20)21/h1-2,5,9-12,14-19H,3-4H2,(H,20,21)/t9-,10-,11+,12-,14+/m0/s1
Formel vikt 330.1