missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).
Solubility
Soluble in methanol.
Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).
Solubility
Soluble in methanol.
Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
Specifications
Melting Point | 206°C to 210°C |
Quantity | 25 g |
Beilstein | 2436216 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 262.05 |
Percent Purity | 98% |
Chemical Name or Material | 3-Iodo-4-methylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'