Läs mer
3-Isopropoxybenzoic Acid, TRC
Produktkod. 30407543
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407543 500 mg
30405135 1 g
30407695 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30407543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI918195500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60772-67-0
Molekylformel C10H12O3
Molekylvikt (g/mol) 180.2
Synonym 3-(1-Methylethoxy)benzoic Acid, 3-Isopropoxybenzoic Acid
IUPAC-namn 3-propan-2-yloxybenzoic acid
LEDER CC(C)Oc1cccc(c1)C(=O)O
Specifikationer
Kemiskt namn eller material 3-Isopropoxybenzoic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C10H12O3/c1-7(2)13-9-5-3-4-8(6-9)10(11)12/h3-7H,1-2H3,(H,11,12)
Formel vikt 180.0786