missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropoxypropylamine, 98+%, Thermo Scientific™

Catalog Number 11440178
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
2906-12-9
MFCD00008220
This item is not returnable. View return policy
2906-12-9
MFCD00008220

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.84
Boiling Point 149°C to 151°C
Flash Point 42°C (108°F)
Molecular Formula C6H5BrFN
Refractive Index 1.4195
Quantity 50 mL
UN Number UN2733
Synonym 5-bromo-2-fluorobenzenamine, 2-fluoro-5-bromoaniline, 5-bromo-2-fluoro aniline, 5-bromo-2-fluorophenylamine, benzenamine, 5-bromo-2-fluoro, 5-bromo-2-fluoro-phenylamine, 1-amino-5-bromo-2-fluorobenzene, 5-bromo-2fluoroaniline, pubchem13482, acmc-1cjje
InChI Key ADWKOCXRCRSMLQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)N)F
IUPAC Name 5-bromo-2-fluoroaniline
Molecular Weight (g/mol) 190.02
PubChem CID 2769407
Formula Weight 117.19
Percent Purity ≥98%
Chemical Name or Material 3-Isopropoxypropylamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'