Läs mer
3-Keto Fusidic Acid, TRC
Produktkod. 30010525
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010525 5 mg -
30003638 2.5 mg 2.5mg
30005932 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK1889805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4680-37-9
Molekylformel C31 H46 O6
Molekylvikt (g/mol) 514.69
Synonym (4alpha,8alpha,9beta,11alpha,13alpha,14beta,16beta,17Z)-16-(Acetyloxy)-11-hydroxy-3-oxo-29-nordammara-17(20),24-dien-21-oic acid, 11alpha,16beta-Dihydroxy-3-oxo-29-nor-8alpha,9beta,13alpha,14beta-dammara-17(20),24-dien-21-oic acid 16-acetate, 3-Oxofusidic acid
IUPAC-namn (2Z)-2-[(4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-11-hydroxy-4,8,10,14-tetramethyl-3-oxo-1,2,4,5,6,7,9,11,12,13,15,16-dodecahydrocyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
LEDER C[C@H]1[C@@H]2CC[C@@]3(C)[C@@H]([C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C)[C@@]2(C)CCC1=O
Specifikationer
Kemiskt namn eller material 3-Ketofusidic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C31H46O6/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32/h9,18,21-22,24-25,27,34H,8,10-16H2,1-7H3,(H,35,36)/b26-20-/t18-,21-,22-,24+,25-,27-,29-,30-,31-/m0/s1
Formel vikt 514.3294
Procent renhet >95%