Läs mer
3-Ketocarbofuran, TRC
Produktkod. 30407166
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30407166 100 mg
1 options
Produktkod. 30407166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK181650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carbofuran-3-keto
CAS 16709-30-1
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H13 N O4
Kvantitet 100 mg
Synonym Carbamic acid, methyl-, ester with 7-hydroxy-2,2-dimethyl-3(2H)-benzofuranone (8CI), 2,2-Dimethyl-7-[[(methylamino)carbonyl]oxy]-3(2H)-benzofuranone, 3(2H)-Benzofuranone, 7-hydroxy-2,2-dimethyl-, methylcarbamate (8CI), 2,2-Dimethyl-3-oxo-2,3-dihydro-1-benzofuran-7-yl methylcarbamate, 3-Ketocarbofuran, 3-Oxocarbofuran
InChI formel InChI=1S/C12H13NO4/c1-12(2)10(14)7-5-4-6-8(9(7)17-12)16-11(15)13-3/h4-6H,1-3H3,(H,13,15)
LEDER CNC(=O)Oc1cccc2C(=O)C(C)(C)Oc12
IUPAC-namn (2,2-dimethyl-3-oxo-1-benzofuran-7-yl) N-methylcarbamate
Molekylvikt (g/mol) 235.24
Formel vikt 235.0845