missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals
1159.76 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, water, moisture.
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, water, moisture.
Specifications
Melting Point | 168°C to 170°C |
Quantity | 250 mg |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 266.21 |
Percent Purity | 99% |
Chemical Name or Material | 3-Maleimidopropionic acid N-hydroxysuccinimide ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'