missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-propanol, 97+%, Thermo Scientific Chemicals

Catalog Number p-3586355
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19721-22-3
C3H8OS
92.16
MFCD00192262
SHLSSLVZXJBVHE-UHFFFAOYSA-N
3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
88211
3-sulfanylpropan-1-ol
C(CO)CS
This item is not returnable. View return policy
19721-22-3
C3H8OS
92.16
MFCD00192262
SHLSSLVZXJBVHE-UHFFFAOYSA-N
3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
88211
3-sulfanylpropan-1-ol
C(CO)CS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.0620g/mL
Boiling Point 106°C (50.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula SH(CH2)3OH
Refractive Index 1.4950 to 1.497
Quantity 1 g
Specific Gravity 1.062
Solubility Information Solubility in water: slightly soluble
Formula Weight 92.16
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Mercapto-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'