Läs mer
3-Methanethiosulfonyl-N,N-dimethylpropionamide, TRC
Produktkod. 30479191
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479191 10 mg
30473555 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479191 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM25900010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methanethiosulfonyl-N,N-dimethylpropionamide
CAS 359436-82-1
Molekylformel C6 H13 N O3 S2
Kvantitet 10 mg
InChI formel InChI=1S/C6H13NO3S2/c1-7(2)6(8)4-5-11-12(3,9)10/h4-5H2,1-3H3
LEDER CN(C)C(=O)CCSS(=O)(=O)C
IUPAC-namn N,N-dimethyl-3-methylsulfonylsulfanylpropanamide
Molekylvikt (g/mol) 211.3
Formel vikt 211.0337