missing translation for 'onlineSavingsMsg'
Läs mer

3-Methoxy-2',4',6',4-tetrahydroxychalcone (>80%), TRC

Produktkod. 30471750
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471750 100 mg
30465410 250 mg
30467267 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30471750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM269613100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 52218-19-6
Molekylformel C16H14O6
Molekylvikt (g/mol) 302.28
Synonym 2',4,4',6'-Tetrahydroxy-3-methoxychalcone, 3-(4-Hydroxy-3-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)-2-propen-1-one
IUPAC-namn 3-(4-hydroxy-3-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)prop-2-en-1-one
LEDER COc1cc(C=CC(=O)c2c(O)cc(O)cc2O)ccc1O

Specifications

Kemiskt namn eller material 3-Methoxy-2',4',6',4-tetrahydroxychalcone
Kvantitet 100 mg
InChI formel InChI=1S/C16H14O6/c1-22-15-6-9(2-4-11(15)18)3-5-12(19)16-13(20)7-10(17)8-14(16)21/h2-8,17-18,20-21H,1H3
Formel vikt 302.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.