missing translation for 'onlineSavingsMsg'
Läs mer

3-Methoxy-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Produktkod. p-7052412
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15438275

Brand: Thermo Scientific Alfa Aesar H33018.MD

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214384-89-0
Molekylformel C9H7F3O2
Molekylvikt (g/mol) 204.148
MDL-nummer MFCD13185677
InChI-nyckel HSPNVFXNJYKWSK-UHFFFAOYSA-N
Synonym 3-methoxy-2-trifluoromethyl benzaldehyde
PubChem CID 73012639
IUPAC-namn 3-methoxy-2-(trifluoromethyl)benzaldehyde
LEDER COC1=CC=CC(=C1C(F)(F)F)C=O

Specifications

Kemiskt namn eller material 3-Methoxy-2-(trifluoromethyl)benzaldehyde
Färg Yellow
Brytningsindex 1.489
Kvantitet 250 mg
Känslighet Air sensitive
Formel vikt 204.15
Procent renhet 97%
Fysisk form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.