Läs mer
3’-Methoxy-4’-nitroflavone, TRC
Produktkod. 30481755
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481755 50 mg
30479265 5 mg
30471517 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26636050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145370-39-4
Molekylformel C16H11NO5
Molekylvikt (g/mol) 297.26
Synonym 2-(3-Methoxy-4-nitrophenyl)-4H-1-benzopyran-4-one
IUPAC-namn 2-(3-methoxy-4-nitrophenyl)chromen-4-one
LEDER O=C1C2=CC=CC=C2OC(C3=CC=C([N+]([O-])=O)C(OC)=C3)=C1
Specifikationer
Kemiskt namn eller material 3'-Methoxy-4'-nitroflavone
Kvantitet 50 mg
InChI formel InChI=1S/C16H11NO5/c1-21-16-8-10(6-7-12(16)17(19)20)15-9-13(18)11-4-2-3-5-14(11)22-15/h2-9H,1H3
Formel vikt 297.0637