Läs mer
3-Methoxy Acetaminophen, TRC
Produktkod. 30012702
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012702 10 mg
30006337 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012702 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22605010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3251-55-6
Molekylformel C9 H11 N O3
Molekylvikt (g/mol) 181.19
Synonym Acetamide, N-(4-hydroxy-3-methoxyphenyl)-, m-Acetanisidide, 4'-hydroxy- (6CI,7CI,8CI), N-(4-Hydroxy-3-methoxyphenyl)acetamide, 3-Methoxyacetaminophen, 3-Methoxy acetaminophen, 3-Methoxyparacetamol, 3-Methoxy paracetamol, 4-Acetamido-2-methoxyphenol, 4-Acetylamino-2-methoxyphenol
IUPAC-namn N-(4-hydroxy-3-methoxyphenyl)acetamide
LEDER COc1cc(NC(=O)C)ccc1O
Specifikationer
Kemiskt namn eller material 3-Methoxy Acetaminophen
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H11NO3/c1-6(11)10-7-3-4-8(12)9(5-7)13-2/h3-5,12H,1-2H3,(H,10,11)
Formel vikt 181.0739
Procent renhet >95%