Läs mer
3-Methoxy Benzopyrene, TRC
Produktkod. 30482313
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30482313 10 mg
1 options
Produktkod. 30482313 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26103510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methoxy Benzopyrene
CAS 63059-68-7
Molekylformel C21H14O
Kvantitet 10 mg
Synonym 3-Methoxy Benzo[a]pyrene
InChI formel InChI=1S/C21H14O/c1-22-19-11-8-13-6-9-17-16-5-3-2-4-14(16)12-15-7-10-18(19)20(13)21(15)17/h2-12H,1H3
LEDER COc1ccc2ccc3c4ccccc4cc5ccc1c2c35
Molekylvikt (g/mol) 282.34