Learn More
3-Methoxy-p-tyramine Hydrochloride, TRC
Product Code. 17903785
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903785 250 mg 250mg
17982675 100 mg 100mg
17914905 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17903785 Supplier Toronto Research Chemicals Supplier No. TRCM332320250MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1477-68-5
Molecular Formula C9 H13 N O2 . Cl H
Molecular Weight (g/mol) 203.67
Synonym Phenol, 4-(2-aminoethyl)-2-methoxy-, hydrochloride (1:1), Guaiacol, 4-(2-aminoethyl)-, hydrochloride (6CI), Phenol, 4-(2-aminoethyl)-2-methoxy-, hydrochloride (8CI,9CI), 2-Methoxytyramine hydrochloride, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 3-O-Methyldopamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride, 4-Hydroxy-3-methoxy-β-phenethylamine hydrochloride, 4-Hydroxy-3-methoxyphenethylamine hydrochloride, 3-Methoxy-p-tyramine hydrochloride
IUPAC Name 4-(2-aminoethyl)-2-methoxyphenol;hydrochloride
SMILES Cl.COc1cc(CCN)ccc1O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H
Formula Weight 203.0713
Percent Purity >95%
Chemical Name or Material 3-Methoxy-p-tyramine Hydrochloride
Grade HPLC