Läs mer
3-Methoxy-propionaldehyde (Technical Grade), TRC
Produktkod. 30469249
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469249 50 mg
30468833 500 mg
30468479 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30469249 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33619350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2806-84-0
Molekylformel C4H8O2
Molekylvikt (g/mol) 88.11
IUPAC-namn 3-methoxypropanal
LEDER COCCC=O
Specifikationer
Kemiskt namn eller material 3-Methoxy-propionaldehyde (Technical Grade)
Kvantitet 50 mg
InChI formel InChI=1S/C4H8O2/c1-6-4-2-3-5/h3H,2,4H2,1H3
Formel vikt 88.0524