missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7021571
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
591-31-1
C8H8O2
136.15
MFCD00003361
WMPDAIZRQDCGFH-UHFFFAOYSA-N
m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
11569
3-methoxybenzaldehyde
COC1=CC=CC(=C1)C=O
This item is not returnable. View return policy
591-31-1
C8H8O2
136.15
MFCD00003361
WMPDAIZRQDCGFH-UHFFFAOYSA-N
m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
11569
3-methoxybenzaldehyde
COC1=CC=CC(=C1)C=O

3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

Solubility
Slightly soluble in water. Soluble in glacial acetic acid, alcohol, chloroform, ether, carbon disulfide, methanol, aqueous solutions of alkali hydroxides.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Density 1.118
Boiling Point 231°C to 233°C
Flash Point 121°C (250°F)
Odor Characteristic
Refractive Index 1.553
Quantity 25 g
Beilstein 606013
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water. Soluble in glacial acetic acid,alcohol,chloroform,ether,carbon disulfide,methanol,aqueous solutions of alkali hydroxides.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'