Learn More
3-Methoxytyramine hydrochloride, MedChemExpress
Product Code. 16451576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
1g
250mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451576 500 mg 500mg
16431029 1 g 1g
16419855 250 mg 250mg
16414926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16451576 Supplier Medchem Express Supplier No. HY103638
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress 3-Methoxytyramine hydrochloride is an inactive metabolite of dopamine which can activate trace amine associated receptor 1 (TAAR1).
3-Methoxytyramine hydrochloride is an inactive metabolite of dopamine which can activate trace amine associated receptor 1 (TAAR1).
Specifications
Chemical Name or Material 3-Methoxytyramine hydrochloride
CAS 1477-68-5
Color Brown
Purity Grade Notes Research
Molecular Formula C9H14ClNO2
Quantity 500 mg
Synonym 3-O-methyl Dopamine hydrochloride
Solubility Information DMSO : 100 mg/mL (490.99 mM; Need ultrasonic and warming)
SMILES COC1=C(O)C=CC(CCN)=C1.Cl
Molecular Weight (g/mol) 203.67
Formula Weight 203.67
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation